Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trifluoroacetic acid ≥99%, BioSyn™ for protein sequencing, purified by redistillation, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: TFA

299537-500G 299537-100G 299537-25G
BJ299537-25GEA 119.02 USD InStock
BJ299537-25G BJ299537-500G BJ299537-100G
Trifluoroacetic acid ≥99%, BioSyn™ for protein sequencing, purified by redistillation, Fluka™
Trifluoroacetic acid


Formula: CF₃COOH
MW: 114.02 g/mol
Boiling Pt: 72 °C (1.013 hPa)
Melting Pt: -15 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg
Order Now
Specification Test Results
Identity (IR) complying
Assay (GC) Min. 99.0 %
Assay 99.0 - 101.0 %
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.05 %
Suitability complying
APHA Max. 25

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty