Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.5 M NaOH (0.5N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

319503-2L 319503-500ML
BJ319503-500MLEA 31.99 USD InStock
BJ319503-500ML BJ319503-2L
Sodium hydroxide 0.5 M NaOH (0.5N) volumetric solution, Fluka™
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: >100 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 1.020 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,III
Order Now
Specification Test Results
Assay 0.4900 - 0.5100 N
Carbonate (as Na₂CO₃) Max. 0.010 N
Traceable to NIST SRM complying
Vapor Density >1 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty