Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥99.9%, CHROMASOLV™ for LC-MS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: 2-HydroxypropaneIPAIsopropanolIsopropyl alcoholsec-PropanolPropan-2-olPropane, 2-hydroxy-sec-Propyl AlcoholRubbing Alcohol

34965-4X4L 34965-1L 34965-2.5L
BJ34965-1LCS 1317.93 USD InStock
BJ34965-1L BJ34965-2.5L BJ34965-4L
2-Propanol ≥99.9%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
2-Propanol
2-Propanol (Isopropyl alcohol, Isopropanol, IPA) is a colorless solvent and is the simplest example of a secondary alcohol. It is moderately polar and has a boiling point of 82 °C. 2-Propanol dissolves a wide range of non-polar compounds and it evaporates quickly. Thus, it is used widely as a cleaning fluid, especially for dissolving oils. Suitable for LC-MS and cleaning LC-MS systems.

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 77-83 °C (1.013 hPa)
Melting Pt: -90 °C
Density: 0.784-0.786 g/cm³ (20 °C) (lit.)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
Particles > 0,5 µm effective P/ml
Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as C₂H₅COOH) Max. 0.001 %
Silver (Ag) Max. 0.1 ppm
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Sodium (Na) Max. 0.1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Transmittance at 220 nm Min. 50 %
Transmittance at 230 nm Min. 75 %
Transmittance from 260 nm Min. 98 %
HPLC Gradient at 254 nm Max. 5 mAU
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
Suitability for the LC-MS complying
Vapor Pressure 44 mmHg ( 25 °C)
Refractive Index n20/D 1.377 (lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty