Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

34966-4X4L 34966-4X2.5L 34966-1L 34966-2.5L
BJ34966-1LEA 194.29 USD InStock
BJ34966-1L BJ34966-4X2.5L BJ34966-4L BJ34966-2.5L
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Methanol
Methanol (CH3OH) is also known as Methyl Alcohol or wood alcohol. It is a clear, colorless, volatile and flammable liquid. Methanol is a common laboratory solvent and is especially useful for HPLC, UV/VIS spectroscopy, due to its low UV cutoff. Methanol is toxic if ingested and can result in blindness, coma and death. Suitable for LC-MS applications and spectrophotometry.

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities


Formula: H₃COH
MW: 32.04 g/mol
Boiling Pt: 64-65 °C (1.013 hPa)
Melting Pt: -98 °C
Density: 0.79 g/cm³ (20 °C) (lit.)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Vapor Density 1.11 (vs air)
Assay (GC) Min. 99.90 %
Non-volatile matter max. 0.0005 %
Water (Karl Fischer) max. 0.02 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as HCOOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0005 %
Silver (Ag) Max. 0.1 ppm
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Sodium (Na) Max. 0.1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.02 ppm
Tin (Sn) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Absorbance at 210 nm Max. 0.523
Absorbance at 220 nm Max. 0.301
Absorbance at 230 nm Max. 0.125
Absorbance at 260 nm Max. 0.009
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
HPLC Gradient at 254 nm Max. 5 mAU
Suitability for the LC-MS complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty