Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥98.0% (by titrimetric analysis), pellets, Puriss. p.a. ACS (max. 0.02% K), Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

71690-500G 71690-1KG 71690-5KG
BJ71690-500GEA 134.13 USD InStock
BJ71690-500G BJ71690-5KG BJ71690-1KG
Sodium hydroxide ≥98.0% (by titrimetric analysis), pellets, Puriss. p.a. ACS (max. 0.02% K), Fluka™
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Assay Min. 98.0 %
Assay of Na₂CO₃ Max. 1.0 %
Silver (Ag) Max. 0.0005 %
Aluminium (Al) Max. 0.0005 %
Barium (Ba) Max. 0.0005 %
Bismuth (Bi) Max. 0.0005 %
Calcium (Ca) Max. 0.0005 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Mercury (Hg) Max. 0.00001 %
Potassium (K) Max. 0.02 %
Lithium (Li) Max. 0.0005 %
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.0005 %
Molybdenum (Mo) Max. 0.0005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Strontium (Sr) Max. 0.0005 %
Thallium (Tl) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Ag) Max. 0.002 %
Chloride (Cl) Max. 0.005 %
Total N Max. 0.0003 %
Phosphate (PO₄) Max. 0.0005 %
Silicate (as SiO₂) Max. 0.0005 %
Sulfate (SO₄) Max. 0.003 %
Vapor Pressure 3 mmHg ( 37 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty