Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trifluoroacetic acid ≥99.0% (by GC), BioSyn™ for biosynthesis, for HPLC, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: TFA

91707-250ML 91707-250ML
BJ91707-250MLEA 1382.49 USD InStock
CABJ91707250ML BJ91707-250ML
Trifluoroacetic acid ≥99.0% (by GC), BioSyn™ for biosynthesis, for HPLC, Burdick & Jackson™
Trifluoroacetic acid


Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 72 °C (1.013 hPa)
Melting Pt: -15 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg
Order Now
Specification Test Results
Refractive Index n20/D 1.3 (lit.)
Assay (GC) Min. 99.0 %
Suitability test (HPLC) complying
Water (Karl Fischer) Max. 0.05 %
Absorbance at 320 nm Max. 0.025
Absorbance at 300 nm Max. 0.03
Absorbance at 290 nm Max. 0.04
Absorbance at 280 nm Max. 0.05
Absorbance at 270 nm Max. 0.1
Absorbance at 260 nm Max. 0.9
APHA Max. 25

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty