Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyridine ≥99.8%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: AzabenzeneAzine

BN333-20 BB333-2.5L BC333-200 BB333-4 BB333-4 BB333-2.5L
BJBC333-200EA 47307.18 USD InStock
BJBC333-200 BJBN333-20 CABJBB333-4 BJBB333-2.5L CABJBB333-2.5L BJBB333-4
Pyridine ≥99.8%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Pyridine
BioSyn« products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity


Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 12.4 %(V)
Melting Pt: -41.55 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 12,08153
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/97/790797.jpg
Order Now
Specification Test Results
Amines by Ninhydrin Test 10 ppm max.
Free Acid Passes test
GC Analysis 99.8 % min.
Refractive Index (20ºC) 1.5085 - 1.5109
Residue 1 mg/L max.
Water by Karl Fischer Titration 50 ppm max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty