Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile, low water content ≥99.9% (by GC), BioSyn™ for biosynthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, cyano-Ethyl nitrileMethyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

BC017-56 BC017-200 BC017-1250
BJBC017-1250EA 30908.2 USD InStock
BJBC017-1250 BJBC017-56 BJBC017-200
Acetonitrile, low water content ≥99.9% (by GC), BioSyn™ for biosynthesis, Burdick & Jackson™
Acetonitrile


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C
Melting Pt: –46 °C
Density: 0.7822 g/cm³
Flash Pt: 6 °C
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
GC Analysis 99.9% min
Refractive Index (20 °C) 1.3434 - 1.3446
Residue Max. 1 mg/l
UV Absorbance at 200 nm Max. 0.050 AU
UV Absorbance at 210 nm Max. 0.040 AU
UV Absorbance at 225 nm Max. 0.010 AU
UV Cutoff Max. 190 nm
Water by Karl Fischer Titration Max. 20 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty