Synonyms:
5-Ethyltio-1H-Tetrazole
SR721-2.5
BC721-56
BR721-4
BR721-2
SR721-1
BR721-20
BR721-2
SR721-1
SR721-1
BR721-4X2.5L
SR721-2.5
BR721-4X2.5L
BR721-4
BC721-200
BJBR721-2CS
60705.6
USD
InStock
BJBR721-2
CABJBR721-4
CABJBR721-2
BJBC721-200
BJBR721-4
BJSR721-2.5
CABJSR721-2.5
CABJSR721-1
BJBR721-20
BJBR721-4X2.5L
BJBC721-56
CABJBR72142.5L
BASR721-1NAN
BJSR721-1
5-(Ethylthio)-1H-tetrazole (ETT) 0.25M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
5-(Ethylthio)-1H-tetrazole (ETT)
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination