Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene ≥99.9%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: MethacideBenzene, methylMethylbenzolToluolPhenylmethaneMethylbenzeneMethane, phenyl-FM approval

BN346-20
BJBN346-20EA 1069.53 USD InStock
BJBN346-20
Toluene ≥99.9%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Toluene


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C
Melting Pt: -95 °C
Density: 0.867 g/cm3
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
UN: 1294
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty