Synonyms:
TCA
, Aceto-Caustin
, Trichloroethanoic acid
BR605-2
BR605-4X2.5L
BR605-2
BR605-20
BR605-4
BR605-4
BR605-4X2.5L
BJBR605-4X2.5LCS
2286.72
USD
InStock
BJBR605-4X2.5L
CABJBR6054X2.5
CABJBR605-4
BJBR605-4
BJBR605-2
CABJBR605-2
BJBR605-20
Trichloroacetic acid (TCA) 3% (w/v) in dichloromethane, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Trichloroacetic acid (TCA)
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
UNSPSC Code: 12350000