Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloroacetic acid (TCA) 3% (w/v) in dichloromethane, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: TCAAceto-CaustinTrichloroethanoic acid

BR605-2 BR605-4X2.5L BR605-2 BR605-20 BR605-4 BR605-4 BR605-4X2.5L
BJBR605-4X2.5LCS 2286.72 USD InStock
BJBR605-4X2.5L CABJBR6054X2.5 CABJBR605-4 BJBR605-4 BJBR605-2 CABJBR605-2 BJBR605-20
Trichloroacetic acid (TCA) 3% (w/v) in dichloromethane, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Trichloroacetic acid (TCA)
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis

  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

UNSPSC Code: 12350000


Formula: Cl₃CCOOH
MW: 163.39 g/mol
Boiling Pt: 40 °C
Melting Pt: -95 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: Does not flash. Closed cup (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
UN: 2922
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/52/727452.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty