BR663-2
BR663-4X2.5L
BR663-2
BR663-4X2.5L
BJBR663-4X2.5LCS
1719.56
USD
InStock
BJBR663-4X2.5L
CABJBR663-2
BJBR663-2
CABJBR6634X2.5
Oxidation reagent 0.02 M I₂ in 78/20/2 THF/pyridine/water (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Iodine
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- Ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
DNA reagents specifically processed and purified for lab scale oligonucleotide synthesis. Suitable for ABI 380 and 390 series and 3948 DNA/RNA synthesizers. Mixture passed through 0.2 µm filter prior to packaging to minimize particulate contamination.