BR665-4
BJBR665-4CS
1912.69
USD
InStock
BJBR665-4
Oxidation reagent 0.02 M I₂ in 89.6/10/0.4 THF/water/pyridine (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Iodine
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- Ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination