Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-(Ethylthio)-1H-tetrazole (ETT) 0.6M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: 5-Ethyltio-1H-Tetrazole

BR726-4 BR726-1 BR726-4
BJBR726-1CS 3885.79 USD InStock
BJBR726-1 BJBR726-4 CABJBR726-4
5-(Ethylthio)-1H-tetrazole (ETT) 0.6M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
5-(Ethylthio)-1H-tetrazole (ETT)
Reagents are specifically processed and purified to ensure the high synthesis efficiency

  • Ideal for oligonucleotide synthesis
  • Extremely low water content
  • High purity reagents
  • Low amine specifications

Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.


Formula: C₃H₆N₄S
MW: 130.17 g/mol
Boiling Pt: 75 °C
Density: 0.8117 g/cm3
Flash Pt: 6.11 °C
Storage Temperature: Ambient
MDL Number: MFCD00068733
CAS Number: 89797-68-2
UN: 1648
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty