Synonyms:
5-Ethyltio-1H-Tetrazole
BR731-20
BR731RN-4
BC731-200
BR731-4
SR731-1
BR731-4
BJBR731-4CS
10748.75
USD
InStock
CABJBR731-4
BJBR731-4
BJBC731-200
BJBR731RN-4
BJBR731-20
BJSR731-1
5-(Ethylthio)-1H-tetrazole (ETT) 0.3M in acetonitrile + 0.5% NMI, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
5-(Ethylthio)-1H-tetrazole (ETT)
Reagents are specifically processed and purified to ensure the high synthesis efficiency
- Ideal for oligonucleotide synthesis
- Extremely low water content
- High purity reagents
- Low amine specifications
Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.