Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.8%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, cyano-Ethyl nitrileMethyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

PP016TC19 PP016-204 PP016-204 PP016TC19
BJPP016-204EA 10646.28 USD InStock
CABJPP016-204 CABJPP016TC19 BJPP016-204 BJPP016TC19
Acetonitrile ≥99.8%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Acetonitrile
For specialty organic synthesis, preparative scale liquid chromatography, glassware cleaning, and applications requiring ACS reagent grade solvent. The 54-gallon drum is individually analyzed with certificate attached.

  • Low residue
  • Purified solvent
  • ACS General Use certified


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C
Melting Pt: -46 °C
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 6 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Appearance Passes test
Assay by GC Analysis 99.9 % min.
Color (APHA) 10 max.
Refractive Index (20ºC) 1.3430 - 1.3450
Residue after Evaporation 5 mg/L max.
Titrable Acid 8 µeq/g max.
Titrable Base 0.6 µeq/g max.
Water by Karl Fischer Titration 0.1 % max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty