Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

S5881-500G S5881-5KG S5881-6X1KG
BJS5881-1KGCS 657.75 USD InStock
BJS5881-1KG BJS5881-5KG BJS5881-500G
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Vapor Density >1 (vs air)
Assay Min. 98.0 %
Assay of Na₂CO₃ Max. 1.0 %
Appearance of the solution complying
Turbidity of solution Max. 3.5 NTU
Vapor Pressure 3 mmHg ( 37 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty