Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloroacetic acid (DCA) 3% (v/v) in dichloromethane, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: DCADCA Deblock

CS622-200 SR622-4 SR622-20
BJCS622-200EA 13063.88 USD InStock
BJCS622-200 BJSR622-4 BJSR622-20
Dichloroacetic acid (DCA) 3% (v/v) in dichloromethane, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Dichloroacetic acid (DCA)
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis

  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 40 °C (main component)
Melting Pt: -95 °C (main component)
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 98.9 °C (main component)
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
EINECS: 201-207-0
UN: 2922
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty