Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SX4750 and SX4750A Swinging Bucket Rotors, Beckman Coulter®

Supplier: Beckman Coulter
393042 369702 393042 369702 369704 369702 369704
BK369704EA 7412.88 USD InStock
CABK393042 BK369704 MPCABK369702 BK369702 CABK369704 BK393042 CABK369702
SX4750 and SX4750A Swinging Bucket Rotors, Beckman Coulter®
Centrifuges Rotors
These rotors are for use with Allegra® X-12, X-12R, X-14, and X-15R centrifuges.

  • Application: Rapid Sediment of Protein Precipitates, Large Particles, Cells, and Cell Debris
  • Rotor Material: Stainless Steel
  • Bucket Material: Anodized Aluminum
  • Maximum Capacity: 750 mL
  • Minimum Capacity: 1.5 mL

They feature sturdy stainless steel yoke construction with free-swinging aluminum buckets

Rotors may be outfitted to accept tubes as small as 1.5 mL and bottles up to 750 mL in volume, cell culture flasks, blood bags, and microplates (model SX4750 only).

The SX4750A unit provides ARIES™ self-balancing technology that automatically corrects imbalances up to 50 g between opposing loads without shutdown. Individual, BioCertified bucket covers minimize contamination from broken or leaking tubes.

Certifications: Covers are certified CAMR, Porton Down, UK for biocontainment.

Ordering information: Rotors are supplied with four buckets. Rotor bucket covers (BK392805) are sold separately and are supplied with gaskets and cover clips. Adapters are available to accommodate a variety of vessels; see BK359481 series (Aerosolve® canisters), BK359148 and BK343108 (tubes), BK349946 (bottles), BK369295 (cell culture flasks), BK356856 (blood bags).

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty