Would you like to log in again?
Biomek® Pipette Tips for Biomek Liquid Handlers, Beckman Coulter®
Supplier: Beckman Coulter
Biomek®
B85940 B01091 B85965 B85894 B01113 B85894 C41861 B85975 B01124 A22288 717253 987925 719012 B85771 C41862 B85749 987936 B85917 B85745 B85760 394622 719222 B85739 B85881 B85903 B85888 B85764 609043 717255 B85922 B85881 719013 B85922 394627 A21578 B85936 A21578 B85872 B85955 B85917 B85915 B85929 C41864 A21582 B85899 394263 B85975 394627 379502 717252 B01114 A22287 B85771 B85760 B85888 B01103 B01102 B85926 B01123 B85753 379504 A22287 A21586 379504 140505 987935 B85915 B01123 B01090 B85961 B85955 719223 987936 719011 A21586 B85965 B01122 B85892 719010 379506 987935 B85906 394622 A22288 717253 B85745 B85892 B85936 B01122 B01124 717251 B01089 B85775 B01112 B85940 B01102 719011 B85961 A21582 B85767 717256 379503 B85767 B85756 379505 379503 C41865 717251 379502 379501 B85877 B85959 B85872 B85959 B85903 B01101 B85911 717254 717252 B85945 B85756 B85775 379505 B85971 B85981 B01090 B85749 717255 987925 719012 B85753 719013 B01101 379506 B01113 B85884 B01103 B85877 394263 B85945 C41863 B01114 B85764 719223 B85884 B85906 B01112 B01089 373687 B85926 B85899 B85981 379501 719222 B85739 B01091 B85929 717254 B85971 B85911 717256
76112-044CS 217.87 USD InStock
CABKB01122 CABKB01089 76112-044 CABKB01123 CABKB01124 76112-042 BKB01101 76112-040 BKB01102 BKB01103 76334-646 CA76112-050 76112-048 76112-046 CA76111-894 CA76111-652 CA76111-892 CA76111-656 CA76111-654 CA76111-896 BK987935 BK987936 CABK719223 CABK719222 CA76111-890 BKA22287 BKA22288 76111-882 76112-034 CA76112-044 76112-032 CA76112-046 CABKB01112 CABKB01113 CA76112-048 BKB01112 76112-030 BKB01113 BKB01114 CABK379503 BK373687 CABK379504 CABK379501 CABK379502 CA76112-040 76112-038 76334-938 CABKB01114 CA76112-042 76334-936 76112-036 CABK379505 CABK379506 CA76111-882 76334-942 76334-940 BK717256 BK987925 BK717255 BK717254 BK717253 BK717252 BK717251 CABKB01101 CABKB01102 CABKA21586 CABKA21582 CABKB01103 76111-664 76111-666 76111-668 CABK394263 BK719222 BK719223 BKA21578 76111-660 76111-662 CABKA22288 76112-050 BK394263 CA76111-658 CABKA22287 76111-894 76111-652 CA76111-664 76111-896 CA76111-662 76111-654 CA76111-668 76111-656 CA76111-666 76111-658 BKA21582 BK609043 CA76111-660 76111-890 76111-892 BKA21586 CA76112-012 CA76112-014 CA76112-016 CA76112-018 CA76112-010 CABKA21578 BK379502 BK379501 BK379504 BK379503 BK379506 BK379505 CABK987925 CABK717251 CABK717252 CABK717253 CABK717254 CABK717255 CABK717256 76112-022 CA76112-034 76112-020 CA76112-036 BKB01123 CA76112-038 BKB01124 CABK987935 CABK987936 CABK394627 BKB01122 BKB01089 76112-028 76112-026 CA76112-030 CA76112-032 76112-024 76111-864 CABK394622 76112-012 CA76112-022 CA76112-024 76112-010 CA76112-026 CA76112-028 BK140505 76112-018 76112-016 76112-014 CA76112-020 BK719010 BKB01090 BK719011 BK394622 BKB01091 BK719012 BK719013 BK394627 CA76111-864 CABKB01091 CABK719012 CABK719011 CABK719013 CABKB01090
Biomek® Pipette Tips for Biomek Liquid Handlers, Beckman Coulter®
Pipette Tips
Biomek® Pipette Tips from Beckman Coulter® are the only tips on the market that are designed, validated, and approved for use on Biomek® Liquid Handlers.
  • Color-coded racks
  • Easy visual checks
Tip definitions, pipetting techniques, and templates are built into Biomek® software. For best in total system performance, use only Beckman Coulter® validated pipette tips on Biomek® workstations.
Products that are designated as Sterile are either sterilized using Gamma radiation or ethylene oxide (EtO) gas in accordance with ANSI/AAMI/150 11135 or ANSI/AAMI/ISO 11137 guidelines. The sterilization processes certify a sterility assurance level of 10-6. Biomek® P30, P30XL, P20, P50, and P250 tips are EtO gas-sterilized for critical low-volume pipetting performance.
Tip racks are color-coded and matched in Biomek® software for easy-to-identify visual checks during method development and confirmation of the deck before a run.
Certifications: All pipette tips are certified to be free from DNase/RNase, DNA (Human and Mouse), PCR inhibition, pyrogen, endotoxins, and trace-metals.
Ordering information: AP384 tips are designed for use on the Biomek® AP384 Multichannel Head. AP96 Tips are designed for use on the Biomek® AP96 Multichannel head and Biomek® 3000 Pipetting Tools. Span-8 tips are designed for use on the Biomek® Span-8 pod. Biomek® 2000 and 3000 tips are designed for use on the respective pipetting tools.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty