Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) oxide ≥97%

Supplier: BeanTown Chemical
Synonyms: Copper monoxide granulatedCopper oxideCupric oxide

136820-2KG 136820-500G 136820-10KG
BT136820-2KGEA 130.84 USD InStock
BT136820-2KG BT136820-500G BT136820-10KG
Copper(II) oxide ≥97%
Copper (II) oxide
  • RTECS: GL7900000
  • -325 Mesh Powder


Formula: CuO
MW: 79.55 g/mol
Melting Pt: 1330 °C
Density: 6.4 g/mL
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty