Would you like to log in again?
Cholesterol Oxidase, MP Biomedicals
Supplier: MP Biomedicals
0215067425 0215067483 0215067480
IC15067480EA 254.15 USD InStock
IC15067480 IC15067483 IC15067425
Cholesterol Oxidase, MP Biomedicals
Enzymes
CHOD is soluble in cold 50 mM potassium phosphate buffer, pH 7.0
One unit causes the formation of one µmole of hydrogen peroxide per minute at pH 7.0 and 37 °C.
Prepare solutions immediately before use.
Stabilizers: Bovine serum albumin, sugars. Typical Contaminants: Catalase: Not more than 1.0% Cholesterol esterase: Not more than 1.0 × 10^2%. Michaelis Constant: 4.3 × 10^-5 M (Cholesterol); Inhibitors: Ionic detergents, Hg2+, Ag+; Optimum Temperature: 45 - 50°C ; Thermal Stability: below 45 °C (pH 7.0, 15 minutes)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty