Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide ≥85.0%, pellets, BAKER ANALYZED®, Electronic Grade, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Potash causticPotassium hydrateCaustic potash

Management of Change (MOC) category = R

3141-01 BAKR3141-01 3141-05 BAKR3141-05 3141-07 BAKR3141-19
JT3141-5CS 950.26 USD InStock
CAJT3141-5 CAJT3141-19 JT3141-5 CAJT3141-1 JT3141-7 JT3141-1
Potassium hydroxide ≥85.0%, pellets, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320 °C (1013 hPa)
Melting Pt: 360 °C
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
REACH: 01-2119487136-33
Merck Index: 12,07806
Order Now
Specification Test Results
For Microelectronic Use
Assay (KOH) ≥85.0 %
Potassium Carbonate (K₂CO₃) ≤1.0 %
Insoluble Matter ≤50 ppm
Chloride (Cl) ≤0.01 %
Sulfate (SO₄) ≤20 ppm
Ammonium Hydroxide Precipitate ≤0.02 %
Sodium (Na) ≤0.05 %
Nitrogen Compounds (as N) ≤10 ppm
Phosphate (PO₄) ≤5 ppm
Heavy Metals (as Ag) ≤10 ppm
Iron (Fe) ≤10 ppm
Mercury (Hg), For Information Only
Nickel (Ni) ≤10 ppm
Calcium (Ca) ≤50 ppm
Magnesium (Mg), For Information Only

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty