Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 45% (w/w) ∼11.7 N, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Potash causticPotassium hydrateCaustic potash

Management of Change (MOC) category = N

BAKR3143-R BAKR3143-01 BAKR3143-07 BAKR3143-07 BAKR3143-01 BAKR3143-03 BAKR3143-03
JT3143-7EA 1242.47 USD InStock
CAJT3143-7 CAJT3143-1 JT3143-7 CAJT3143-3 JT3143-R JT3143-3 JT3143-1
Potassium hydroxide 45% (w/w) ∼11.7 N, BAKER ANALYZED®, J.T.Baker®
Potassium hydroxide
Lot analysis on label.


Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (KOH) 45.0 - 46.5 %
Potassium Carbonate (K₂CO₃) ≤0.2 %
Chloride (Cl) ≤0.003 %
Sulfate (SO₄) ≤0.001 %
Ammonium Hydroxide Precipitate ≤0.005 %
Sodium (Na) ≤0.02 %
Nitrogen Compounds (as N) ≤5 ppm
Phosphate (PO₄) ≤2 ppm
Heavy Metals (as Ag) ≤5 ppm
Trace Impurities - Copper (Cu) ≤0.050 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Trace Impurities - Nickel (Ni) ≤5.000 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty