Synonyms:
Potash caustic
, Potassium hydrate
, Caustic potash
Management of Change (MOC) category = N
BAKR3143-R
BAKR3143-01
BAKR3143-07
BAKR3143-07
BAKR3143-01
BAKR3143-03
BAKR3143-03
JT3143-7EA
1242.47
USD
InStock
CAJT3143-7
CAJT3143-1
JT3143-7
CAJT3143-3
JT3143-R
JT3143-3
JT3143-1
Potassium hydroxide 45% (w/w) ∼11.7 N, BAKER ANALYZED®, J.T.Baker®
Potassium hydroxide
Lot analysis on label.