Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 0.1 N in methanol, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Potash causticPotassium hydrateCaustic potash

Management of Change (MOC) category = N

BAKR5650-03 BAKR5650-02 BAKR5650-03 BAKR5650-02
JT5650-3CS 860.13 USD InStock
CAJT5650-2 CAJT5650-3 JT5650-3 JT5650-2
Potassium hydroxide 0.1 N in methanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Density: 0.80 g/cm³
Flash Pt: 12.00 °C
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8),II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Chloride (Cl) ≤10 ppm
Nitrogen Compounds (as N) ≤1 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty