Would you like to log in again?
KONTES® Separatory Funnels, Squibb, with Glass Plug Stopcock, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KONTES®
636000-0500 636010-2000 636000-0250 636000-0060 636010-0060 636010-0250 636000-1000 636000-2000 636000-0125 636000-0030
KT636000-0250EA 260.95 USD InStock
KT636000-0250 KT636000-2000 KT636000-0030 KT636010-2000 KT636010-0250 KT636000-1000 KT636010-0060 KT636000-0125 KT636000-0500 KT636000-0060
KONTES® Separatory Funnels, Squibb, with Glass Plug Stopcock, Kimble Chase, DWK Life Sciences
Funnels Separating Funnels
Squibb-type funnels feature a [ST] joint and a [ST] glass plug stopcock
Plain stem. Available with or without a [ST] stopper.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty