Would you like to log in again?
KIMBLE® Separatory Funnels, Short Stem, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
636040-2000
KT636040-2000EA 754.87 USD InStock
KT636040-2000
KIMBLE® Separatory Funnels, Short Stem, DWK Life Sciences
Funnels Separating Funnels
These separatory funnels are designed for rapid extractions of environmental samples.
  • Funnel top has a short drip tip and a standard taper 34/28 opening for more convenient filling
  • Stopper is PTFE
  • Thick walls help resist breakage in the cleaning and drying cycle
  • Manufactured from 33 expansion, low extractable borosilicate glass
PTFE stopcock plug is KT821001-0006 and has a 6 mm bore for rapid transfer.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty