Would you like to log in again?
CPC® Quick-Disconnect Fittings, Hose Barb Bodies
Supplier: CPC (Colder Products Company)
PLCD17004 (25PK) PLCD1600412 PMCD1703 (1/PK) PMCD1704 (25PK) PMC1604 PMCD1603 (25PK) PMCD1603 (1/PK) PMCD160112 (25PK) PLCD17005 (25PK) PMCD1701 (1/PK) PMC1702 (25PK) PLCD1600412 (25PK) PMC1601 PLC17006 (1/PK) PLCD1700612 (25PK) PLC17005 PMCD1703 (25PK) PMC1603 PMC1703 (1/PK) PMCD1602 (25PK) PMCD1602 (1/PK) PMCD170112 (25PK) PMC1601 (25PK) PLCD1600612 PMCD160412 PMC1701 PMCD1604 (25PK) PLCD16005 (25PK) PMC1602 PMCD160112 PLCD16006 (25PK) PMC1703 (25PK) PLCD16004 (25PK) PMCD160212 (25PK) PLCD1700612 PMCD170412 (25PK) PLCD1700412 PLCD17004 (1/PK) PLC16004 (25PK) PMCD160212 PLC16006 (1/PK) PMCD160412 (25PK) PLC17006 (25PK) PMCD1601 (25PK) PMCD1702 (25PK) PMCD1704 (1/PK) PMCD170112 PMCD1701 (25PK) PLC17004 (1/PK) PLCD17006 (25PK) PMCD170212 PMCD1702 (1/PK) PMC1602 (25PK) PLC17005 PLC16004 (1/PK) PMCD170412 PMCD1604 (1/PK) PLCD16006 (1/PK) PLC17004 (25PK) PLC16006 (25PK) PMCD1601 (1/PK) PLCD16005 (1/PK) PLCD1700412 (25PK) PMCD170212 (25PK) PMC1704 PLCD17006 (1/PK) PMC1702 (1/PK) PLCD17005 (1/PK) PLCD1600612 (25PK) PLCD16004 (1/PK) PMC1603 (25PK)
MFLX06360-10EA 22.8 USD InStock
MFLX06360-10 MFLX06364-65BLK MFLX06360-55 MFLX06361-51BLK MFLX06364-19 MFLX06360-15 MFLX06360-12 MFLX31307-04 MFLX06361-03BLK MFLX06364-15 MFLX06360-57 MFLX06364-10 MFLX06364-55 MFLX06360-10BLK MFLX06360-65BLK MFLX06364-55BLK MFLX06361-12BLK MFLX06360-19 MFLX06361-12 MFLX06361-11 MFLX06360-55BLK MFLX06361-53 MFLX06361-52BLK MFLX06364-70BLK MFLX06361-04BLK MFLX06361-13 MFLX06364-10BLK MFLX06361-52 MFLX06361-51 MFLX06360-20BLK MFLX06364-25BLK MFLX06361-11BLK MFLX06361-46BLK MFLX06364-09 MFLX06360-09 MFLX06364-15BLK MFLX06361-53BLK MFLX06361-05 MFLX06364-19BLK MFLX06361-04 MFLX06361-48 MFLX06361-03 MFLX06361-05BLK MFLX06361-46 MFLX06364-70 MFLX06360-12BLK MFLX06360-70 MFLX06360-67BLK MFLX06364-09BLK MFLX06361-09 MFLX06360-25BLK MFLX06364-60BLK MFLX06364-20BLK MFLX06360-65 MFLX06360-22 MFLX06360-57BLK MFLX06360-20 MFLX06360-25 MFLX06364-25 MFLX06360-67 MFLX06364-20 MFLX06364-65 MFLX06364-60 MFLX06360-15BLK MFLX06360-60 MFLX06360-19BLK MFLX06360-70BLK MFLX06360-60BLK MFLX06360-09BLK MFLX06361-48BLK MFLX06360-22BLK
CPC® Quick-Disconnect Fittings, Hose Barb Bodies
Fittings and Connectors
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty