Would you like to log in again?
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Santoprene®, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
6431-12 6428-42 6431-42 6431-26
MFLX06431-42EA 84 USD InStock
MFLX06431-42 MFLX06431-12 MFLX06428-42 MFLX06431-26
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Santoprene®, Avantor®
Tubing Peristaltic Pump Tubing
Choose the size and formulation to suit your flow rate and application needs.
  • Genuine Ismatec® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Compatible with Masterflex® L/S® Multichannel pump head, small cartridges
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty