Would you like to log in again?
Masterflex® L/S® Standard Pump Heads for Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
7018-21 7014-20 7017-20 7013-20 7014-21 7014-52 7016-52 7013-21 7016-20 7018-20 7013-52 7017-52 7017-21 7016-21 7018-52
MFLX07013-20CS 0 USD InStock
MFLX07013-20 MFLX07014-21 MFLX07014-20 MFLX07013-52 MFLX07013-21 MFLX07016-20 MFLX07017-21 MFLX07016-21 MFLX07018-21 MFLX07017-20 MFLX07014-52 MFLX07016-52 MFLX07018-20 MFLX07017-52 MFLX07018-52
Masterflex® L/S® Standard Pump Heads for Precision Tubing, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Ideal for fluid transfer applications requiring high accuracy and repeatability.
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Stack up to four heads depending on drive recommendations
This Standard pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 17, L/S 18.
Delivery information: Includes tubing loading key and single-channel mounting hardware.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty