Would you like to log in again?
CPC® AseptiQuik® G Series Genderless Aseptic Connectors
Supplier: CPC (Colder Products Company)
CPC® AseptiQuik® G
AQG17112 AQG17MPCHT(5PK) AQG17108 AQG17006 AQG17008HT AQG17004 AQG17104 AQG17MPC AQG17106 AQG17006HT
MFLX20202-36EA 75.4 USD InStock
MFLX20202-36 MFLX20202-57 MFLX20202-56 MFLX20202-45 MFLX20202-55 MFLX20202-54 MFLX20202-43 MFLX20202-42 MFLX20202-40 MFLX20202-38
CPC® AseptiQuik® G Series Genderless Aseptic Connectors
Fittings and Connectors
Provide easy, secure sterile connections even in nonsterile environments.
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use hose barb connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.
Simply fit the connector half in your tubing line. The easy three-step FLIP-CLICK-PULL assembly procedure ensures a tight seal with no additional hardware required. The audible “click” confirms a securely connected assembly.
All fittings can be sterilized by gamma irradiation. Use high-temperature fittings to sterilize via autoclave.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty