Would you like to log in again?
Masterflex® Union Fittings, Hose Barb, Tee, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
T0-2-200AGPP T0-2-200PP (25 PACK) T0-1.5-200PP(10 PACK) T0-5PP(100 PACK) T0-12 AGPP 227945 T0-1.5-200PP(250 PACK) T0-16PP-MF(10PK) T0-10PP-MF(10PK) T0-4PP-MF(10PK) T0-6PP-MF(10PK) T0-5PP(10 PACK) T0-3PP(50 PACK) T0-2-200PP-MF (100 PACK) T0-10PP(10 PACK) T0-8PP(10 PACK) T0-2-200PP (250 PACK) T0-12PP-MF(10PK) T0-16PP(5 PACK) T0-8PP-MF(10PK) 227925 227965 T0-1-200PP-MF(10PK) T0-10PP(100/PK) T0-12PP(50/PK) T0-6PP(100/PK) CEA33-PR0 T0-8PP(100/PK) 227955 T0-3PP(25 PACK) T0-1.5-200PP(25 PACK) T0-3PP-MF(10PK) T0-8 AGPP T0-1-200PP (10 PACK) T0-6PP(10 PACK) T0-2-200PP-MF(10PK) 227935 227915 T0-4PP(10 PACK) 227905 T0-4PP(100/PK) T0-12PP(5 PACK) T0-4 AGPP T0-1.5-200PP(100 PACK)
MFLX50624-30EA 23.6 USD InStock
MFLX50624-30 MFLX50623-66 MFLX06294-40 MFLX50627-66 MFLX06294-60 MFLX06294-20 MFLX06368-84 MFLX06368-82 MFLX16368-81 MFLX40610-30 MFLX40610-31 MFLX30613-63 MFLX40613-43 MFLX40610-34 MFLX40613-48 MFLX40610-35 MFLX30613-67 MFLX40610-32 MFLX40613-47 MFLX40610-33 MFLX50627-57 MFLX40627-76 MFLX40623-70 MFLX06294-50 MFLX06294-70 MFLX40623-76 MFLX50624-28 MFLX06294-10 MFLX50623-57 MFLX06294-30 MFLX06365-77 MFLX50628-29 MFLX06368-85 MFLX40640-33 MFLX40640-34 MFLX30613-51 MFLX30613-71 MFLX30613-55 MFLX40640-30 MFLX40640-31 MFLX30613-75 MFLX40640-32 MFLX30613-59 MFLX40627-70
Masterflex® Union Fittings, Hose Barb, Tee, Polypropylene, Avantor®
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty