Would you like to log in again?
Masterflex® Reducer Fittings, Hose Barb, Straight (Nylon), Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
C2.5-2NN(10 PACK) C3-2BN(10PK) C2-1.5-200BN(10PK) C1.5-1-200NN(250 PACK) C4.2-5NN(10 PACK) C5-4NN(10 PACK) C4-3NN(100 PACK) C2.5-2BN(10PK) C2-1.5-200NN(250 PACK) C10-8NN-MF(10PK) C8-6-NN(10 PACK) C16-10NN(5 PACK) C12-8NN-MF(10PK) C5-4NN(100 PACK) C4-2BN(10PK) C10-6NN(10 PACK) C8-6-NN(100 PACK) C2-1-200NN(250 PACK) C3-2NN-MF(10PK) C10-8NN(10 PACK) C10-4NN(10 PACK) C4-3NN(10 PACK) C0-1-200BN(10PK) C12-10NN(50 PACK) C8-5NN(10 PACK) C4-2NN(10 PACK) C10-5NN(10 PACK) C2-1-200NN-MF(10PK) C8-5NN(100 PACK) C12-6NN(5 PACK) C1.5-1-200NN(25 PACK) C4-2.5BN(10PK) C6-4NN(10 PACKS) C2-1.5-200NN-MF(10PK) C2-1-200BN(10PK) C16-8NN(5 PACK) C16-8NN(50 PACK) C4-2NN(100 PACK) C12-6NN(50 PACK) C12-8NN(50 PACK) C8-4NN(100 PACK) C2-1.5-200NN(25 PACK) C16-12NN(5 PACK) C8-4NN(10 PACK) C10-8NN(100 PACK) C6-4NN(100 PACK) C12-10NN(5 PACK) C5-4BN(10PK) C2-1-200NN(25 PACK) C12-8NN(5 PACK)
MFLX31518-35EA 25.5 USD InStock
MFLX31518-35 MFLX31518-32 MFLX30626-10 MFLX41518-30 MFLX30622-10 MFLX30622-12 MFLX30626-16 MFLX30622-14 MFLX31518-39 MFLX31518-37 MFLX30622-16 MFLX30616-10 MFLX30622-08 MFLX30626-28 MFLX30622-25 MFLX31518-27 MFLX30622-28 MFLX30622-18 MFLX40616-02 MFLX30621-99 MFLX30621-97 MFLX30616-26 MFLX30703-99 MFLX40616-06 MFLX30626-30 MFLX30622-30 MFLX30626-34 MFLX30622-34 MFLX30626-39 MFLX30622-39 MFLX30616-30 MFLX40626-20 MFLX40625-94 MFLX30626-41 MFLX40621-94 MFLX30626-43 MFLX30622-41 MFLX40626-22 MFLX40622-20 MFLX30622-43 MFLX30626-04 MFLX40622-22 MFLX30626-06 MFLX30622-02 MFLX30626-08 MFLX30622-04 MFLX41518-28 MFLX30622-06 MFLX41517-92 MFLX30704-89
Masterflex® Reducer Fittings, Hose Barb, Straight (Nylon), Avantor®
Fittings and Connectors
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperature range −50 to +250 °F (−46 to +121 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty