Would you like to log in again?
Masterflex® Reducer Fittings, Male NPT Threaded, Straight, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
RN6-2NK7-MF 10PK RN4-2NK7-MF 10PK MAKE-UP RN6-2PP(10 PACK) RN8-6NN(60 PACK) RN6-4K(10 PACK) RN6-4NN(6 PACK) RN8-2NK7-MF 10PK RN8-2PP(10 PACK) RN8-2NN(6 PACK) RN8-6PP(100 PACK) MPT-3412B-10 RN6-2NN(6 PACK) RN6-2HDPE(10 PACK) RN8-6PP(10 PACK) RN4-2HDPE(10 PACK) RN8-4HDPE(10 PACK) RN6-4PP(100 PACK) RN8-6NN(6 PACK) RN8-4NK7-MF 10PK RN8-6NK7-MF 10PK MPT-3412B-100 RN4-2NN(6 PACK) RN8-4PP(10 PACK) MAKE-UP RN4-2PP(10 PACK) RN4-2PP(100 PACK) RN6-4PP(10 PACK) RN6-4HDPE(10 PACK) RN8-4NN(6 PACK) RN8-6HDPE(10 PACK) MAKE-UP
MFLX40627-43EA 99.6 USD InStock
MFLX40627-43 MFLX30623-40 MFLX30623-41 MFLX30623-42 MFLX40623-43 MFLX30623-46 MFLX40623-47 MFLX30623-48 MFLX06349-86 MFLX06349-87 MFLX40627-55 MFLX30627-54 MFLX40623-51 MFLX30623-50 MFLX30623-52 MFLX40623-55 MFLX40627-35 MFLX30623-32 MFLX30623-54 MFLX40623-35 MFLX30623-34 MFLX30623-36 MFLX40623-39 MFLX30623-38 MFLX31902-00 MFLX40615-33 MFLX40615-34 MFLX40615-35 MFLX40615-31 MFLX31701-88 MFLX40615-32 MFLX31701-89
Masterflex® Reducer Fittings, Male NPT Threaded, Straight, Avantor®
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty