Would you like to log in again?
CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Inserts
Supplier: CPC (Colder Products Company)
HFCD221012 (1/PK) HFCD221012 (10PK) HFC221012 (1/PK) HFCD22635 (10PK) HFCD22635 (1/PK) HFCD22835 (1/PK) HFC22635 (10PK) HFCD221235 (10PK) HFC22835 (10PK) HFC22635 (1/PK) HFC22812 (10PK) HFCD221235 (1/PK) HFCD221212 (1/PK) HFC22812 (1/PK) HFCD22835 (10PK) HFCD22612 (10PK) HFC221212 (1/PK) HFCD22812 (1/PK) HFC221235 HFC22612 (10PK) HFC22835 (1/PK) HFC221212 (10PK) HFCD22812 (10PK) HFCD221035 HFCD221212 (10PK) HFC22612 (1/PK) HFCD22612 (1/PK) HFC221012 (10PK)
MFLX31303-46EA 20.2 USD InStock
MFLX31303-46 MFLX31303-47 MFLX31303-48 MFLX31307-46BLK MFLX31303-42 MFLX31307-47 MFLX31307-46 MFLX31303-41BLK MFLX31303-43 MFLX31307-40BLK MFLX31307-48 MFLX31303-45 MFLX31307-45 MFLX31303-40 MFLX31303-41 MFLX31307-48BLK MFLX31308-74 MFLX31307-41 MFLX31307-40 MFLX31303-47BLK MFLX31303-45BLK MFLX31303-43BLK MFLX31307-45BLK MFLX31303-40BLK MFLX31307-41BLK MFLX31303-48BLK MFLX31303-46BLK MFLX31303-42BLK
CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Inserts
Fittings and Connectors
No more leaks—valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty