Would you like to log in again?
Masterflex® Electro-Optic Point-Level Sensors, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
187585 157750 143575 143585 194472 143590 194471 224505 187580 153063 143580
MFLX32772-08EA 273 USD InStock
MFLX32772-08 MFLX68964-02 MFLX32772-04 MFLX32772-02 MFLX32772-12 MFLX32772-00 MFLX32772-22 MFLX32772-10 MFLX32772-32 MFLX32772-20 MFLX32772-30
Masterflex® Electro-Optic Point-Level Sensors, Avantor®
Level Sensors
No clog or wear—contain no moving parts.
  • Detect liquids regardless of conductivity, color, or specific gravity
  • Economical and compact — built-in solid-state switching electronics
  • One-unit installation — simply screws into the top or side of your tank
These dependable, electro-optic level sensors contain no moving parts to clog or wear. Sensors offer ±1 mm repeatability and are available in a wide variety of materials to meet even the most demanding applications.
Installation is simple and quick through the tank top, bottom, or side. The switches can be used for low-, intermediate-, or high-liquid level detection. Solid state switching ensures dependability over a long service life. Order controller separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty