Would you like to log in again?
Masterflex® Plastic Assorted Fitting Kits, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
JK139BN JK202HDPE JK201BN EJK138PP KIT PP EJK140BN EJ203HDPE(10PK) EJK203PP EJK200PP JK201HDPE JK200BN JK138HDPE KIT HDPE EJK140PP JK202BN JK141BN EJK137PP EJK141PP JK200HDPE JK137BN JK141HDPE JK137HDPE EJK138NG KIT, NK EJK202PP EJK201-PP EJK140HDPE JK139HDPE EJK203-BN EJK139PP EJK138NG KIT, BN
MFLX41514-30EA 157 USD InStock
MFLX41514-30 MFLX41514-11 MFLX31514-32 MFLX31514-10 MFLX31514-17 MFLX41514-19 MFLX31514-18 MFLX41514-35 MFLX31514-33 MFLX31514-34 MFLX31514-13 MFLX41514-15 MFLX31514-14 MFLX31514-36 MFLX31514-09 MFLX31514-21 MFLX31514-26 MFLX41514-27 MFLX31514-05 MFLX31514-06 MFLX41514-07 MFLX31514-29 MFLX31514-22 MFLX31514-01 MFLX41514-23 MFLX31514-02 MFLX41514-03 MFLX31514-25
Masterflex® Plastic Assorted Fitting Kits, Avantor®
Fittings and Connectors
Get organized today!
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Assorted fittings come in an impact-resistant plastic box with secure snap-shut latches to prevent spillage
  • Labeled compartments let you easily locate the fittings you want
  • Order storage racks separately
Organize your workstation with these convenient fitting kits. Assorted fittings come in impact resistant plastic box with secure latch to keep fittings organized and in place. Label compartments make fitting location quick and easy. Choose from four materials: high-density polyethylene (HDPE), black nylon, natural polypropylene, and natural PVDF.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty