Would you like to log in again?
Monoject™Syringes, Nonsterile, Covidien
Supplier: Covidien
Monoject™
8881112083 8881103025 8881120029 8881112059 8881135084 8881106028
82017-954CS 189.54 USD InStock
82017-954 82017-644 MJ8881-120029 MJ8881-103025 15141-128 15141-126
Monoject™Syringes, Nonsterile, Covidien
Syringes Syringes, Medical
These latex-free syringes feature standard graduations and can be autoclaved or gas sterilized
Bulk packed.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty