Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Methyl Isobutyl KetoneIsobutyl Methyl KetoneMethyl iso-Butyl KetoneMIK

Management of Change (MOC) category = N

MACR6247-04 MACR6247-19 MACR6247-08 MACR6247-08 MACR6247-19 MACR6247-04
MK624719EA 927.32 USD InStock
CAMK624708 CAMK624719 MK624719 MK624708 CAMK624704 MK624704
4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
4-Methyl-2-pentanone


Formula: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Boiling Pt: 117.4 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.801 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://us-legacy.vwr.com/stibo/web/std.lang.all/93/40/739340.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water) ≥98.5 %
ACS - Color (APHA) ≤15
ACS - Residue after Evaporation ≤0.005 %
ACS - Titrable Acid (meq/g) ≤0.002
ACS - Water (H₂O) ≤0.1 %
Appearance (clear, colorless liquid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty