Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide ≥85%, pellets, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Potash causticPotassium hydrateCaustic potash

Management of Change (MOC) category = N

MACR6984-28 MACR6984-28 MACR6984-12 MACR6984-69 MACR6984-20 MACR6984-06 6984-04 MACR6984-20 MACR6984-04
MK698404CS 829.1 USD InStock
CAMK698420 MK698404 MK698428 MK698406 CAMK698404 CAMK698428 MK698420 MK6984-69 MK698412
Potassium hydroxide ≥85%, pellets, AR® ACS, Macron Fine Chemicals™
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320 °C (1013 hPa)
Melting Pt: 360 °C
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
REACH: 01-2119487136-33
Merck Index: 12,07806
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KOH) ≥85 %
Identification Passes Test
Calcium (Ca) ≤0.005 %
Chloride (Cl) ≤0.01 %
Heavy Metals (as Ag) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Nickel (Ni) ≤0.001 %
ACS - Magnesium (Mg) ≤0.002 %
Nitrogen Compounds (as N) ≤0.001 %
ACS - Phosphate (PO₄) ≤5 ppm
Sodium (Na) ≤0.05 %
Potassium Carbonate (K₂CO₃) ≤2.0 %
ACS - Sulfate (SO₄) ≤0.003 %
Appearance (white hygroscopic pellets)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty