Would you like to log in again?
Aluminium Sample Pan for Tzero Sample Press Kit, TA Instruments
Supplier: TA INSTRUMENTS INC MS
901683.901
MSPP-901683901PK 476.92 USD InStock
MSPP-901683901
Aluminium Sample Pan for Tzero Sample Press Kit, TA Instruments
Trays/Dishes/Pots Instrument Trays/Dishes
This aluminum pan is designed for maximum performance in crimp or hermetic sealing of various sample sizes.
  • Temperature Range: –180 to 600 °C
Flat bottom design for maximum contact with the Tzero cell sensors.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty