Would you like to log in again?
GoTaq MDx Hot Start Polymerase, Promega
Supplier: Promega
GoTaq®
D6005 D6006 D6001 D6008
PAD6008EA 5126.01 USD InStock
PAD6008 PAD6006 PAD6005 PAD6001
GoTaq MDx Hot Start Polymerase, Promega
Nucleic Acid Reagents End-point PCR Enzymes and Kits
GoTaq MDx Hot Start Polymerase contains GoTaq MDx DNA Polymerase bound to a proprietary antibody that blocks polymerase activity.
  • Inhibits Polymerase Activity at Temperatures below 70°C
  • Manufactured under cGMP
  • High-concentration and glycerol-free formulations also available
  • All formulations supplied at ≥5u/microl, except for high-concentration formulation, which is supplied at ≥50u/microl
  • Custom formats and formulations
GoTaq MDx Hot Start Polymerase contains GoTaq MDx DNA Polymerase bound to a proprietary antibody that blocks polymerase activity. The polymerase activity is restored during the initial denaturation step when amplification reactions are heated at 94-95°C for two minutes. This allows hot-start PCR in which polymerase activity is inhibited at temperatures below 70°C, allowing convenient, room-temperature reaction setup. Hot-start PCR is advantageous for some amplification targets because primer-dimer and secondary products are eliminated or minimized. In some cases, hot-start PCR may improve yields. The glycerol-free product formulation is further purified to remove glycerol, making it suitable for further manufacture processing and lyophilization. GoTaq MDx DNA Polymerase products are manufactured under cGMP. GoTaq MDx DNA Polymerase products are general purpose reagents intended for general laboratory use. They can be used as a component of molecular diagnostic assays, where applicable country laws allow, without paying royalties. The products by themselves do not provide any diagnostic result.
The glycerol-free product formulation is further purified to remove glycerol, making it suitable for further manufacture processing and lyophilization. GoTaq MDx DNA Polymerase products are manufactured under cGMP standards.
The polymerase activity is restored during the initial denaturation step when amplification reactions are heated at 94 to 95°C for two minutes. This allows hot-start PCR, in which polymerase activity is inhibited at temperatures below 70°C. Hot-start PCR is advantageous for some amplification targets because primer-dimer and secondary products are eliminated or minimized. In some cases, hot-start PCR may improve yields.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty