Would you like to log in again?
DeadEnd Colorimetric TUNEL System, Promega
Supplier: Promega
G3250 G7130 G7360
PAG7130EA 802.16 USD InStock
PAG7130 PAG7360 PAG3250
DeadEnd Colorimetric TUNEL System, Promega
Assays Cellular Assays
A modified TUNEL Assay designed for simple, rapid detection of apoptotic cells in situ at the single-cell level. Measures nuclear DNA fragmentation.
  • A Modified TUNEL Assay for Detection of Apoptotic Cells In Situ
  • Detects apoptosis in tissue sections or cell cultures
The DeadEnd Colorimetric TUNEL System is a modified TUNEL Assay designed to provide simple, accurate and rapid detection of apoptotic cells in situ at the single-cell level. The DeadEnd Colorimetric TUNEL System measures nuclear DNA fragmentation, an important biochemical indicator of apoptosis. The system can be used to assay apoptotic cell death in both tissue sections and cultured cells. The DeadEnd Colorimetric TUNEL System end-labels the fragmented DNA of apoptotic cells using a modified TUNEL (TdT-mediated dUTP Nick-End Labeling) assay. Biotinylated nucleotide is incorporated at the 3'-OH DNA ends using the enzyme Terminal Deoxynucleotidyl Transferase (TdT). Horseradish-peroxidase-labeled streptavidin (Streptavidin HRP) is then bound to these biotinylated nucleotides, which are detected using the peroxidase substrate, hydrogen peroxide, and the stable chromogen, diaminobenzidine (DAB). Using this procedure, apoptotic nuclei are stained dark brown.
Using this procedure, apoptotic nuclei are stained dark brown. The protocol for the DeadEnd TUNEL Assay recommends an optional DNase I treatment of samples as a positive control to detect DNA fragmentation.
Biotinylated nucleotide is incorporated at the 3´-OH DNA ends using Terminal Deoxynucleotidyl Transferase (TdT). Horseradish-peroxidase-labeled streptavidin (Streptavidin HRP) is then bound to these biotinylated nucleotides, which are detected using the peroxidase substrate, hydrogen peroxide, and the stable chromogen, diaminobenzidine (DAB).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty