Would you like to log in again?
GoTaq® G2 Green Master Mix, 1000 reactions, Promega
Supplier: Promega
GoTaq®
M7822 M7823
PAM7822EA 48.94 USD InStock
PAM7822 PAM7823
GoTaq® G2 Green Master Mix, 1000 reactions, Promega
Nucleic Acid Reagents End-point PCR Enzymes and Kits
GoTaq(R) G2 Master Mixes are premixed ready-to-use solutions containing GoTaq(R) G2 DNA Polymerase, dNTPs, MgCl2 and reaction buffers at optimal concentrations for efficient amplification of a wide range of DNA templates by PCR.
  • All-in-One PCR Master Mix: Just Add Template and Primers
  • High-Performance GoTaq(R) G2 DNA Polymerase in a Ready-to-Use Master Mix
  • Simplifies reaction setup and saves time
  • Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
GoTaq G2 Green and Colorless Master Mixes are ready-to-use, green and colorless, 2X master mixes containing all necessary components for robust, reliable PCR, including the second-generation GoTaq G2 DNA Polymerase. Add template, primers and go. The second generation of GoTaq products, GoTaq G2 DNA Polymerase reliably amplifies a wide range of PCR templates and provides high-performing results due to improved manufacturing processes, increased reliability and consistency.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty