Would you like to log in again?
Pierce™ Immobilized Papain (Agarose Resin), Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
20341 20341
PI20341EA 399.68 USD InStock
PI20341 CAPI20341
Pierce™ Immobilized Papain (Agarose Resin), Thermo Scientific
Protein Extraction Protein Extraction Reagents
Thermo Scientific Pierce Immobilized Papain consists of a cysteine-endopeptidase (papain) that has been immobilized onto beaded agarose resin to enable generation and purification of Fab fragments from antibodies.
  • Papain can also be used to generate F(ab')2 fragments
  • Immobilized papain canbe substituted fro free papain because it eliminates autolysis and contamination of a sample with the protease
  • Allows control of the digestion by removing the pepsin or controlling the flow of sample over an immobilized papain column
  • Immobilized papain is more stable against heat-induced denaturation, resulting in longer maintenance of activity
Papain is a nonspecific, cysteine-endopeptidase isolated from papaya latex that is used in a wide variety of applications. When IgG molecules are incubated with papain in the presence of cysteine, one or more peptide bonds in the hinge region are split, producing three fragments of similar size: two Fab fragments and one Fc fragment. The Fc fragments can be separated from the Fab fragments using immobilized Protein A or by ion exchange chromatography.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty