Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 10 g/L in mixed alcohol solvent for determination of salts in crude oil

Supplier: Ricca Chemical
Danger
Synonyms: HaliteSea saltNaClSaline

R7212000-1C
RC7212-32EA 126.91 USD InStock
RC7212-32
Sodium chloride 10 g/L in mixed alcohol solvent for determination of salts in crude oil
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: ~ 118 °C
Density: 0.8632 g/cm³
Flash Pt: ~ 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
UN: 1987
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear, colorless, Butanol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty