Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
R7375000-500A
R7375000-20F
R7375000-1A
R7375000-4A
RCR7375000500AEA
31.41
USD
InStock
RCR7375000500A
RC73751
RCR7375000-1A
RC73755
Sodium hydroxide 12.5 g/L 0.313 N suitable for crude fiber analysis of food and feed
Sodium hydroxide
AOAC, for crude fiber.