Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK1 Rabbit Monoclonal Antibody [clone: 3G2]

Supplier: Rockland Immunochemicals
100-401-218 100-401-218
RL100-401-218EA 518 USD InStock
RL100-401-218 CA100-401-218
Anti-MAPK1 Rabbit Monoclonal Antibody [clone: 3G2]
Antibodies
This antiserum was tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a predominant band approximately 42 kDa in size corresponding to p42 MAP Kinase (ERK2) by western blotting.

Recommended Dilutions: ELISA: 1:2000 to 1:10000; Western Blot: 1:1000-1:,2000; Immunoprecipitation: 5.0 µg/mg sample;

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 3G2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty